Learn More
MSC Vikan UST Bench Brush, 13 inch, Medium Bristle, Yellow, Polyester/Polypropylene
Catalog No. 502835951
Encompass
The Vikan UST Bench Brush is a 13 inch yellow bench brush with medium bristle stiffness, constructed from Polyester (PBT) and polypropylene. It's designed for effective removal of damp and wet particles from conveyor belts in high-risk areas, utilizing Ultra Bristle Security to minimize contamination and bristle loss.
  • Ultra safe technology (UST) with ultra bristle security for effective removal of damp and wet particles.
  • Unique bristle system minimizes contamination and bristle loss.
  • Durable construction: polyester (PBT), glass-reinforced, and polypropylene.
  • Complies with major food contact regulations: (EC) 1935/2004, EU 2023/2006/EC, FDA CFR 21, UK 2019 No. 704.
  • Meets REACH regulation (EC) No. 1907/2006.
  • Complies with California proposition 65, Halal, and Kosher.
  • No intentionally added PFAS.
Catalog No. 50-283-5951 Supplier MSC Supplier No. 54799325
The Vikan UST Bench Brush is a 13 inch yellow bench brush with medium bristle stiffness, constructed from Polyester (PBT) and polypropylene. It's designed for effective removal of damp and wet particles from conveyor belts in high-risk areas, utilizing Ultra Bristle Security to minimize contamination and bristle loss.
  • Ultra safe technology (UST) with ultra bristle security for effective removal of damp and wet particles.
  • Unique bristle system minimizes contamination and bristle loss.
  • Durable construction: polyester (PBT), glass-reinforced, and polypropylene.
  • Complies with major food contact regulations: (EC) 1935/2004, EU 2023/2006/EC, FDA CFR 21, UK 2019 No. 704.
  • Meets REACH regulation (EC) No. 1907/2006.
  • Complies with California proposition 65, Halal, and Kosher.
  • No intentionally added PFAS.