Learn More
MSC Vikan Bench Brush, UST, 13 in, Polyester, Medium
Catalog No. 502837066
Encompass
This UST Bench Brush is engineered for effective removal of damp and wet particles, especially from conveyor belts in high-risk areas. Its unique bristle system minimizes contamination and bristle loss, making it ideal for environments requiring stringent hygiene and durability.
  • Designed for effective removal of damp and wet particles.
  • Unique bristle system minimizes contamination and bristle loss.
  • Medium bristle stiffness.
  • Suitable for food contact, complying with EC 1935/2004, FDA CFR 21, and UK 2019 No. 704 regulations.
  • Manufactured under Good Manufacturing Practices (GMP).
  • Meets REACH Regulation (EC) No. 1907/2006.
  • Complies with California Proposition 65, Halal, and Kosher standards.
  • Max usage temperature for food contact: 176 °F.
  • Min usage temperature: -4 °F.
  • Resistant to chemicals with pH values from 2 to 10.5.
Catalog No. 50-283-7066 Supplier MSC Supplier No. 54799309
This UST Bench Brush is engineered for effective removal of damp and wet particles, especially from conveyor belts in high-risk areas. Its unique bristle system minimizes contamination and bristle loss, making it ideal for environments requiring stringent hygiene and durability.
  • Designed for effective removal of damp and wet particles.
  • Unique bristle system minimizes contamination and bristle loss.
  • Medium bristle stiffness.
  • Suitable for food contact, complying with EC 1935/2004, FDA CFR 21, and UK 2019 No. 704 regulations.
  • Manufactured under Good Manufacturing Practices (GMP).
  • Meets REACH Regulation (EC) No. 1907/2006.
  • Complies with California Proposition 65, Halal, and Kosher standards.
  • Max usage temperature for food contact: 176 °F.
  • Min usage temperature: -4 °F.
  • Resistant to chemicals with pH values from 2 to 10.5.