Learn More
Grainger Bench Drill Press 1/3 hp 1/2 Chuck
Catalog No. 502766735
Encompass_Preferred
The 10" Bench Drill Press by Belle Group is a 1/3 HP, 120V drill press with a belt drive system. It has a cast iron head and table, adjustable depth stop, and adjustable table tilt. The spindle travel is 3-1/8" and it has a spindle taper of MT#2. The 16 speeds range from 200-3,630 RPM. The drill press has a swing of 10" and a distance between the spindle and column of 7-7/8". It comes with a key chuck and arbor. The overall dimensions are 15-3/4" W x 27-1/2" D x 37-3/4" H and it weighs 79 lbs.
Catalog No. 50-276-6735 Supplier Grainger Supplier No. 16N196
The 10" Bench Drill Press by Belle Group is a 1/3 HP, 120V drill press with a belt drive system. It has a cast iron head and table, adjustable depth stop, and adjustable table tilt. The spindle travel is 3-1/8" and it has a spindle taper of MT#2. The 16 speeds range from 200-3,630 RPM. The drill press has a swing of 10" and a distance between the spindle and column of 7-7/8". It comes with a key chuck and arbor. The overall dimensions are 15-3/4" W x 27-1/2" D x 37-3/4" H and it weighs 79 lbs.