Learn More
Grainger Bench Grinder 6 36 Grit 1/3HP
Catalog No. 502765611
Encompass_Preferred
The Dayton Bench Grinder Buffer Fixed is a 6 inch bench grinder and buffer that runs at 1725 RPM. It has a 1/3 HP, 120V, 60 Hz motor and 6 inch x 1 inch coarse and medium aluminum oxide grinding wheels. The grinder also includes an adjustable tool rest, eye shields, and rubber feet. This grinder is ideal for sharpening tools and deburring metal. It provides the power and versatility needed for a variety of grinding, sharpening, and buffing applications.
Catalog No. 50-276-5611 Supplier Grainger Supplier No. 16W005
The Dayton Bench Grinder Buffer Fixed is a 6 inch bench grinder and buffer that runs at 1725 RPM. It has a 1/3 HP, 120V, 60 Hz motor and 6 inch x 1 inch coarse and medium aluminum oxide grinding wheels. The grinder also includes an adjustable tool rest, eye shields, and rubber feet. This grinder is ideal for sharpening tools and deburring metal. It provides the power and versatility needed for a variety of grinding, sharpening, and buffing applications.