Learn More
IW Tremont Co Inc LabExact Bench Protector Paper, spill containment, 48cm X 60cm, 100 Sheets
SDP
Catalog No. 502237870
Encompass_Preferred
LabExact Disposable, highly absorbent pure cellulose paper with polyethylene backing is designed to protect laboratory surfaces against liquid spills and prevent leakage through to the bench top, necessary to maintain a safer work environment. Meets OSHA Regulation 29CFR 1910.1030 for Occupational Exposure to Bloodborne Pathogens. Tear resistant material, whether wet or dry. The polyethylene backing material is skid resistant, waterproof and chemical resistant. The absorbent paper quickly traps spilled or drained fluids. Smooth white surface is easy to write on. Sheets are 48x 60cm. Pack of 100 sheets.
This item is not returnable. View return policy
Catalog No. 50-223-7870 Supplier IW Tremont Co Inc Supplier No. LEBPS4657
LabExact Disposable, highly absorbent pure cellulose paper with polyethylene backing is designed to protect laboratory surfaces against liquid spills and prevent leakage through to the bench top, necessary to maintain a safer work environment. Meets OSHA Regulation 29CFR 1910.1030 for Occupational Exposure to Bloodborne Pathogens. Tear resistant material, whether wet or dry. The polyethylene backing material is skid resistant, waterproof and chemical resistant. The absorbent paper quickly traps spilled or drained fluids. Smooth white surface is easy to write on. Sheets are 48x 60cm. Pack of 100 sheets.