Learn More
Accuflow Systems Saint-Gobain Bench Protector, FEP, Vinyl, 25 in x 5 yd
Catalog No. NC2327713
Encompass_Preferred
Saint-Gobain Bytac FEP film bench protector with vinyl backing provides a chemically inert, non-stick protective surface for laboratory benchtops, fume hoods, and work surfaces. The conformable roll adheres to most surfaces, resists common laboratory chemicals, and is easy to cut, apply, clean, and replace.
  • FEP film overlay for chemical resistance and non-stick performance.
  • Vinyl backing conforms to minor surface irregularities.
  • Size: 25 in x 5 yd roll.
  • Easy to cut, apply, clean, and replace.
  • Suitable for benchtops, fume hoods, and tables.
Catalog No. NC2327713 Supplier Accuflow Systems Supplier No. D1069324
Saint-Gobain Bytac FEP film bench protector with vinyl backing provides a chemically inert, non-stick protective surface for laboratory benchtops, fume hoods, and work surfaces. The conformable roll adheres to most surfaces, resists common laboratory chemicals, and is easy to cut, apply, clean, and replace.
  • FEP film overlay for chemical resistance and non-stick performance.
  • Vinyl backing conforms to minor surface irregularities.
  • Size: 25 in x 5 yd roll.
  • Easy to cut, apply, clean, and replace.
  • Suitable for benchtops, fume hoods, and tables.