Learn More
IW Tremont Co Inc LabExact Chemically resistant, highly absorbent green quilted bench pad/wipe, approx. 15x9 inch, 500/cs
SDP
Catalog No. 502237891
Encompass_Preferred
LabExact Use as a bench pad or large wipe. Designed with a complex proprietary fused 100% Polypropylene (PP) fibrous matrix which is a lightweight polyolefin and has an incredibly high chemical resistance. 15 x 9" bench pads or wipes are 290g/m2 basis weight sufficient to easily handle typical lab bench volume spills up to 300ml safely and quickly. Chemically resistant to most short term exposure of organic solvents as well as acids and alkalis. Lays flat as a bench pad with sufficient stiffness reducing ¿bunch up¿ seen with other competing brands. Soft enough to be used as a wipe. Binding dots accumulate fluids and migrate deeper into the pad allowing fast surface absorption and higher fluid hold saturation. Pack of 500.
This item is not returnable. View return policy
Catalog No. 50-223-7891 Supplier IW Tremont Co Inc Supplier No. LEHAP150900CS
LabExact Use as a bench pad or large wipe. Designed with a complex proprietary fused 100% Polypropylene (PP) fibrous matrix which is a lightweight polyolefin and has an incredibly high chemical resistance. 15 x 9" bench pads or wipes are 290g/m2 basis weight sufficient to easily handle typical lab bench volume spills up to 300ml safely and quickly. Chemically resistant to most short term exposure of organic solvents as well as acids and alkalis. Lays flat as a bench pad with sufficient stiffness reducing ¿bunch up¿ seen with other competing brands. Soft enough to be used as a wipe. Binding dots accumulate fluids and migrate deeper into the pad allowing fast surface absorption and higher fluid hold saturation. Pack of 500.