Learn More
IW Tremont Co Inc LabExact Chemically resistant, highly absorbent green quilted bench wipe, approx. 5x4.5 inch, 500/cs
SDP
Catalog No. 502237892
Encompass_Preferred
LabExact Wipes are designed with a complex proprietary fused 100% Polypropylene (PP) fibrous matrix which is a lightweight polyolefin and has an incredibly high chemical resistance. 5 x 4.5" wipes are 290g/m2 basis weight sufficient to easily handle typical small lab bench volume spills safely and quickly. Chemically resistant to most short term exposure of organic solvents as well as acids and alkalis. Lays flat as a small bench pad, soft enough to be used as a wipe. Binding dots accumulate fluids and migrate deeper into the pad allowing fast surface absorption and higher fluid hold saturation. Pack of 500.
This item is not returnable. View return policy
Catalog No. 50-223-7892 Supplier IW Tremont Co Inc Supplier No. LEHAP050450CS
LabExact Wipes are designed with a complex proprietary fused 100% Polypropylene (PP) fibrous matrix which is a lightweight polyolefin and has an incredibly high chemical resistance. 5 x 4.5" wipes are 290g/m2 basis weight sufficient to easily handle typical small lab bench volume spills safely and quickly. Chemically resistant to most short term exposure of organic solvents as well as acids and alkalis. Lays flat as a small bench pad, soft enough to be used as a wipe. Binding dots accumulate fluids and migrate deeper into the pad allowing fast surface absorption and higher fluid hold saturation. Pack of 500.