Learn More
Andwin Scientific Freestanding Bench Rack Hanging System, Gray
SDP
Catalog No. NC9502378
Encompass_Preferred
Bench Rack for AkroBins, Gray. 70.5L x 20.3W x 53.3cm H. For freestanding use at workstations. Bench racks are finished with a durable gray baked enamel finish. Bin fits securely onto panel via full-length hanger lip.
Catalog No. NC9502378 Supplier Andwin Scientific Supplier No. 62661498
Bench Rack for AkroBins, Gray. 70.5L x 20.3W x 53.3cm H. For freestanding use at workstations. Bench racks are finished with a durable gray baked enamel finish. Bin fits securely onto panel via full-length hanger lip.