Learn More
Cole-Parmer A&D Weighing HW-60KC High Resolution Bench Scale; 60 kg x 0.005 kg
Catalog No. NC1743293 Shop All Cole-Parmer Products
Encompass
The HV-C/CP and HW-C/CP bench scales feature a bright 3-5-7 segment color (red-green-yellow) comparator function for checkweighing. A 10-digit keypad makes entering comparator limits and tare weights quick and easy, while a key lock function prevents operators from making unnecessary setting changes. The negative checkweighing function allows you to easily move parts from a master carton on the scale to containers on the production line. Weighing modes include parts counting and accumulation function for totaling weigh data. Weighing units include kg, lb, and oz.
Catalog No. NC1743293 Supplier Cole-Parmer Supplier No. 1112458
The HV-C/CP and HW-C/CP bench scales feature a bright 3-5-7 segment color (red-green-yellow) comparator function for checkweighing. A 10-digit keypad makes entering comparator limits and tare weights quick and easy, while a key lock function prevents operators from making unnecessary setting changes. The negative checkweighing function allows you to easily move parts from a master carton on the scale to containers on the production line. Weighing modes include parts counting and accumulation function for totaling weigh data. Weighing units include kg, lb, and oz.