Learn More
Cole-Parmer Cole-Parmer Shelf Grip Clamp; Nickel-Plated Zinc
Supplier:  Cole-Parmer 0805000
Encompass_Preferred
Designed to affix to a bench or shelf up to 2" (51mm) thick, and attach via the integrated connector a standard 1⁄2" (1.3cm) clamp support rod. The maximum load for this clamp is 150lbs (68kg).
Catalog No. NC1261902