Learn More
Electron Microscopy Sciences 6-Place Table Top Centrifuge, 120 V
SDP
Catalog No. 501899854
Encompass_Preferred
This is a powerful, quiet, centrifuge for table top use in small labs or offices. Features variable 30-minute timer, standard looking lid, and a six place rotor. 7, 10, or 15 mL tube capacity without the added expense of extra tube carriers.
Catalog No. 50-189-9854 Supplier Electron Microscopy Sciences Supplier No. 7235906
This is a powerful, quiet, centrifuge for table top use in small labs or offices. Features variable 30-minute timer, standard looking lid, and a six place rotor. 7, 10, or 15 mL tube capacity without the added expense of extra tube carriers.