Learn More
Research Products International Corp Biocooler Bench-Top Cooler, 1.5 - 2.0ml Tubes, Holds 48
SDP
Catalog No. 50212678
Encompass
Biocooler is designed to consistently maintain subzero temperatures of micro-tubes. Benchtop freezer keeps enzymes and valuable samples at -20°C to -15°C for up to 6 hours at room temperature. Protects samples from temperature fluctuation due to frost-free cycles, power loss, or open freezer doors. Features inner note panel for referencing samples and carrying handle for convenient transporting.
Catalog No. 50-212-678 Supplier Research Products International Corp Supplier No. 260100
Biocooler is designed to consistently maintain subzero temperatures of micro-tubes. Benchtop freezer keeps enzymes and valuable samples at -20°C to -15°C for up to 6 hours at room temperature. Protects samples from temperature fluctuation due to frost-free cycles, power loss, or open freezer doors. Features inner note panel for referencing samples and carrying handle for convenient transporting.