Learn More
Bellco Glass, Inc. Bench Top Digital Roller Apparatus, 230V
SDP
Catalog No. 501945888
Encompass_Preferred
The Bellco Digital Bench-top Roller Apparatus holds 10 glass or plastic roller bottles (110 to 120 mm diameter and up to 22.5 cm in length). The unit features a heavy-duty DC motor and solid-state electronics that operate at bottle speeds ranging from 0.30 to 10.0 rpm with the ability to select clockwise or counter clockwise direction. The digital display panel has a 0.55” LED for easy reading and the apparatus has a locking speed knob. Rollers are easily removed for replacement by using the special tool provided with each unit. All-Position Drive facilitates the use of lightweight plastic roller bottles by ensuring rotation at each bottle station, independent of any load configurations. Bellco’s unique hex-shaped shafts prevent roller slippage for positive drive. Use of this equipment is limited to conventional laboratory mixing of roller bottles. MADE IN THE USA
Catalog No. 50-194-5888 Supplier Bellco Glass, Inc. Supplier No. 762120230
The Bellco Digital Bench-top Roller Apparatus holds 10 glass or plastic roller bottles (110 to 120 mm diameter and up to 22.5 cm in length). The unit features a heavy-duty DC motor and solid-state electronics that operate at bottle speeds ranging from 0.30 to 10.0 rpm with the ability to select clockwise or counter clockwise direction. The digital display panel has a 0.55” LED for easy reading and the apparatus has a locking speed knob. Rollers are easily removed for replacement by using the special tool provided with each unit. All-Position Drive facilitates the use of lightweight plastic roller bottles by ensuring rotation at each bottle station, independent of any load configurations. Bellco’s unique hex-shaped shafts prevent roller slippage for positive drive. Use of this equipment is limited to conventional laboratory mixing of roller bottles. MADE IN THE USA