Learn More
Electron Microscopy Sciences Lab Aire Bag of 19 Small pegs and 6 plugs
SDP
Catalog No. 501902066
Encompass_Preferred
Use with Electric Benchtop Dryers. Redirect air flow by inserting plugs instead of pegs into unused peg holes.
Catalog No. 50-190-2066 Supplier Electron Microscopy Sciences Supplier No. 6440210S
Use with Electric Benchtop Dryers. Redirect air flow by inserting plugs instead of pegs into unused peg holes.