Learn More
Electron Microscopy Sciences Top-Loading Dispensing Bins - Large
SDP
Supplier:  Electron Microscopy Sciences 6438818
Encompass_Preferred
Bins are made from 1/4" thick PVC. They are chemical resistant and easy to clean. Ideal for pipette tips, small vials, cassette lids, cassette base molds, cotton swabs, aluminum stubs, embedding capsules etc.
Footprint: 18 x 10"
Height: 7-3/4"
Catalog No. 50-189-9162