Learn More
Electron Microscopy Sciences Bench Top Mat Cut Size Sheets 20"x24"
SDP
Catalog No. 5024666
Encompass_Preferred
A flexible polyethylene film backing which has a highly absorbent upper surface which traps water, dyes, resins, solvents, and radioactive solutions. The sheets can be rolled up and discarded after use. Ideal for daily use in EM labs. Cut size of 20"x24" sheets.
Catalog No. 50-246-66 Supplier Electron Microscopy Sciences Supplier No. 64446
A flexible polyethylene film backing which has a highly absorbent upper surface which traps water, dyes, resins, solvents, and radioactive solutions. The sheets can be rolled up and discarded after use. Ideal for daily use in EM labs. Cut size of 20"x24" sheets.