Learn More
Andwin Scientific BENCH UNDERPADS
SDP
Catalog No. NC9450069
Encompass
The Deluxe Absorbent Underpads have a three-layer, cushioned construction that is breathable yet absorbent enough. This product is ideal for hospital and laboratory use. The top sheet is made with soft white spunbound polypropylene, while its back sh
Catalog No. NC9450069 Supplier Andwin Scientific Supplier No. 56616030
The Deluxe Absorbent Underpads have a three-layer, cushioned construction that is breathable yet absorbent enough. This product is ideal for hospital and laboratory use. The top sheet is made with soft white spunbound polypropylene, while its back sh