Learn More
Research Products International Corp BenchBin Benchtop Starter Kit
SDP
Catalog No. 502252973
Encompass
BenchBin is a personal benchtop container for biohazard and contaminated waste. Compact size fits perfectly anywhere and safely disposes up to tips, tubes, gloves, etc. Features easy push open/close lid that is perfect for quick disposal of biological waste in compact 7 x 5 x 7” unit. Bin features cover that is easily removed from the inner basin for quick emptying of contents. Ideal size for use with red imprinted autoclavable biohazard bags (8.5x11”, sold separately). BenchBin Starter kit includes one BenchBin and 400 red biohazard bags.
Compact size
Safely disposes tips, tubes, gloves
Easy open lid
This item is not returnable. View return policy
Catalog No. 50-225-2973 Supplier Research Products International Corp Supplier No. 248520
BenchBin is a personal benchtop container for biohazard and contaminated waste. Compact size fits perfectly anywhere and safely disposes up to tips, tubes, gloves, etc. Features easy push open/close lid that is perfect for quick disposal of biological waste in compact 7 x 5 x 7” unit. Bin features cover that is easily removed from the inner basin for quick emptying of contents. Ideal size for use with red imprinted autoclavable biohazard bags (8.5x11”, sold separately). BenchBin Starter kit includes one BenchBin and 400 red biohazard bags.
Compact size
Safely disposes tips, tubes, gloves
Easy open lid