Learn More
MTC Bio BenchBin™ Benchtop Biohazard Bin, 1.5L, 7 x 5 x7in, 1/ea
Catalog No. NC2096021
Encompass_Preferred
BenchBin™ is a personal waste container that any researcher can benefit from. Use with 8.5 x 11inch autoclave bags to dispose of biological waste for subsequent processing or forget the autoclave bag and use the BenchBin as a waste container for non-hazardous waste. The red cover is easily removed from the inner basin for easy emptying of contents. Perfect for pipette, tips, microcentrifuge tubes, and gloves, the BenchBin features a push to open/close lid that keeps materials covered and your bench neat. Safely dispose of tips, tubes, gloves, etc. Secures and covers hazardous waste. Easy push open/close spring lid. Compact footprint – 7 x 5 x 7in.
This item is not returnable. View return policy
Catalog No. NC2096021 Supplier MTC Bio Supplier No. A8401B
BenchBin™ is a personal waste container that any researcher can benefit from. Use with 8.5 x 11inch autoclave bags to dispose of biological waste for subsequent processing or forget the autoclave bag and use the BenchBin as a waste container for non-hazardous waste. The red cover is easily removed from the inner basin for easy emptying of contents. Perfect for pipette, tips, microcentrifuge tubes, and gloves, the BenchBin features a push to open/close lid that keeps materials covered and your bench neat. Safely dispose of tips, tubes, gloves, etc. Secures and covers hazardous waste. Easy push open/close spring lid. Compact footprint – 7 x 5 x 7in.