Learn More
Research Products International Corp Bench Blotter Platform Rocker
SDP
Catalog No. 50550292
Encompass
The BenchBlotter Platform Rocker provides the optimum speed and rocking motion for Western Blotting, gel staining and hybridization applications. Compact low profile design available with an optional dual platform that doubles capacity and saves workspace. Designed for easy portability from the lab bench to a cold room or incubator.
Supplied with one tray, flat non-skid rubber and a two year warranty.
Catalog No. 50-550-292 Supplier Research Products International Corp Supplier No. 248555
The BenchBlotter Platform Rocker provides the optimum speed and rocking motion for Western Blotting, gel staining and hybridization applications. Compact low profile design available with an optional dual platform that doubles capacity and saves workspace. Designed for easy portability from the lab bench to a cold room or incubator.
Supplied with one tray, flat non-skid rubber and a two year warranty.