Learn More
LabScientific Fixed Speed Rocker with non-slip rubber mat, 115V. Optimized for blot washing and gel staining. Feautures a 12X12" surface area. It genty mixes and is both cold room and incubator safe.
Catalog No. NC0015252
Encompass
The rocker (BR1000) is an economical "workhorse" rocker, designed for common molecular lab applications such as blotting, staining, destaining, etc. The controls are easy to follow with no settings, switches, or adjustements to be made. Just press the power switch to begin rocking! Specs: 12 rpm, +/- 8° tilt, 5lbs load capaicty, 6.5lbs weight, 50W power, 12X12" platform area. Dual platform accessory doubles work surface.
Catalog No. NC0015252 Supplier LabScientific Supplier No. BR1000
The rocker (BR1000) is an economical "workhorse" rocker, designed for common molecular lab applications such as blotting, staining, destaining, etc. The controls are easy to follow with no settings, switches, or adjustements to be made. Just press the power switch to begin rocking! Specs: 12 rpm, +/- 8° tilt, 5lbs load capaicty, 6.5lbs weight, 50W power, 12X12" platform area. Dual platform accessory doubles work surface.