Learn More
Sigma Aldrich Fine Chemicals Biosciences BenchBlotterTM 2D platform1EA
Catalog No. 502905887
Encompass
The BenchBlotter is an economical workhorse rocker designed for common molecular lab applications suchas blotting staining/destaining etc. Optimized for blot washing and gel staining the BenchBlotter gently mixes at 12 rpm with a /- 8 tilt. The dual platform accessory doubles the work surface and the unit is cold room and incubator safe.
Catalog No. 50-290-5887 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7554191EA
The BenchBlotter is an economical workhorse rocker designed for common molecular lab applications suchas blotting staining/destaining etc. Optimized for blot washing and gel staining the BenchBlotter gently mixes at 12 rpm with a /- 8 tilt. The dual platform accessory doubles the work surface and the unit is cold room and incubator safe.