Description
- Filter paper with plastic backing
- Apply a few drops of reagent to the paper, then apply it to the suspected bullet hole
- Materials Includes:
- Plastic-backed filter paper
- Options and Accessories:
- FS471: Bullet Hole Testing (BTK) Master Kit
- FS473: Lead Solvent Ampoule Refill
- FS474: Lead Reagent Ampoule Refill
- FS475: Copper Solvent Ampoule Refill
- FS476: Copper Reagent Ampoule Refill
Specifications
Specifications
| Product Type | Test Paper |
| Quantity | 25/Pk. |
| Material | Plastic-backed Filter Paper |