Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GeminiBio BenchMark™ Fetal Bovine Serum
SDP

Catalog No. 11852137 Shop All Gemini Bio Products
Change view
Click to view available options
Gamma-irradiated:
No
Yes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Gamma-irradiated
11-852-137 Yes
11-852-136 No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-852-137 Supplier GeminiBio Supplier No. 100106G500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Extensively characterized serum from USDA-approved sources for reliable, consistent research cell culture.

  • Sourced exclusively from countries of negligible TSE/BSE risk, collected at USDA-approved facilities in countries including the United States, Mexico, and Costa Rica
  • Triple 0.1 μm sterile-filtered for exceptional purity
  • Mycoplasma-tested and virus-screened to 9 CFR 113.53(c) specifications
  • Characterized with an extensive biochemistry panel; low levels of endotoxin and IgG
  • Available standard or gamma-irradiated
  • Up to 5-year shelf life from date of manufacture
  • Ships on dry ice; store frozen at -20°C to -10°C
  • Ideal for tissue culture, stem cell research, biotechnology, and life science research
  • Manufactured under an ISO 13485:2016 certified quality system
  • Complimentary heat inactivation upon request

Specifications

Additional Assays Extensive biochemistry panel, Electrophoretic profile
Certifications/Compliance ISO 13485:2016 certified quality system
Content And Storage 500 mL bottle, store frozen at -20°C to -10°C
Species Bovine
Age Fetal
Country of Origin Countries meeting USDA importation requirements
Form Liquid
Format Bottle
Product Type Fetal Bovine Serum
Quantity 500 mL
Serum Treatment Gamma-irradiated
Sterility Sterile
Sterilization Method Triple-filtered, 0.1 μm, Gamma-irradiated
Source Fetal bovine blood
Grade Premium
Processing Triple 0.1 μm sterile-filtered, gamma-irradiated
Tested For Mycoplasma, Sterility (bacteria and fungi), Adventitious viruses per 9 CFR 113.53(c)
Formulation Liquid, frozen
For Use With (Application) Cell Culture
Heat Inactivated No
IgG levels Low
Lyophilized No
Product Line BenchMark™
Mycoplasma Detected No
Shipping Condition Dry Ice
USDA Safety Testing Included Yes
Regulatory Status RUO
Gamma-irradiated Yes
Show More Show Less

For cell culture, further manufacturing or research use only. Not for direct therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.