Learn More
Benchmark Products Inc BENCHMARK PRODUCTS INC Frame, Touchless, Primary Duty, Non-Sterile, Black
SDP
Catalog No. NC3981541
Encompass
These durable replacement frames are designed for secure mop pad placement. They feature easy-to-replace nylon removable strips for consistent cleaning and maintain touchless operation in cleanroom environments, ensuring continued efficiency and hygiene during disinfecting. They are ideal for walls, ceilings, and floors in sterile or controlled spaces.
  • Secure mop pad placement
  • Easy-to-replace nylon removable strips for consistent cleaning
  • Maintains touchless operation in cleanroom environments
  • Ensures continued efficiency and hygiene during disinfecting
  • Suitable for walls, ceilings, and floors in sterile or controlled spaces
  • Non-sterile
  • Size: 14.75 in. x 0.87 in.
  • Color: Black
Catalog No. NC3981541 Supplier Benchmark Products Inc Supplier No. Y1051650
These durable replacement frames are designed for secure mop pad placement. They feature easy-to-replace nylon removable strips for consistent cleaning and maintain touchless operation in cleanroom environments, ensuring continued efficiency and hygiene during disinfecting. They are ideal for walls, ceilings, and floors in sterile or controlled spaces.
  • Secure mop pad placement
  • Easy-to-replace nylon removable strips for consistent cleaning
  • Maintains touchless operation in cleanroom environments
  • Ensures continued efficiency and hygiene during disinfecting
  • Suitable for walls, ceilings, and floors in sterile or controlled spaces
  • Non-sterile
  • Size: 14.75 in. x 0.87 in.
  • Color: Black