Learn More
Benchmark Products Inc BENCHMARK PRODUCTS INC Telescopic Handle, Anodized Aluminum, 100 to 180 cm, Autoclavable
SDP
Catalog No. NC3981545
Encompass
This telescopic handle is constructed from durable, high-quality anodized aluminum and plastic, designed for comfort and extended use. It is adjustable from 100 cm to 180 cm to suit various user needs and features an anti-slip top grip for enhanced safety. The ergonomic design helps reduce fatigue, and its autoclavable material ensures suitability for various environments.
  • Durable, high-quality anodized aluminum and plastic construction.
  • Adjustable length from 100 cm to 180 cm.
  • Anti-slip top grip to prevent slipping on walls.
  • Color-coded strips for easy tracking of handle locations.
  • Ergonomic grip with rounded ends and soft areas to reduce hand and finger fatigue.
  • Features an anti-flip function.
  • Improved locking mechanism.
  • Lightweight design.
  • Autoclavable material.
Catalog No. NC3981545 Supplier Benchmark Products Inc Supplier No. Y9966M
This telescopic handle is constructed from durable, high-quality anodized aluminum and plastic, designed for comfort and extended use. It is adjustable from 100 cm to 180 cm to suit various user needs and features an anti-slip top grip for enhanced safety. The ergonomic design helps reduce fatigue, and its autoclavable material ensures suitability for various environments.
  • Durable, high-quality anodized aluminum and plastic construction.
  • Adjustable length from 100 cm to 180 cm.
  • Anti-slip top grip to prevent slipping on walls.
  • Color-coded strips for easy tracking of handle locations.
  • Ergonomic grip with rounded ends and soft areas to reduce hand and finger fatigue.
  • Features an anti-flip function.
  • Improved locking mechanism.
  • Lightweight design.
  • Autoclavable material.