Learn More
Cole-Parmer Benchmark Scientific Beadbug 6 Microtube Homogenizer, 115 VAC
Catalog No. NC2038461 Shop All Cole-Parmer Products
Encompass
The BeadBug 6 homogenizer uses a three-dimensional shaking motion. This motion results in high velocity impacts between the sample and the beads. This method is efficient for releasing cellular contents, such as nucleic acids and proteins. The digital display is simple to understand; program options include set speed, time, number of cycles, and pause between cycles. The last programmed parameters are stored in memory so you can easily do repeat runs.
Catalog No. NC2038461 Supplier Cole-Parmer Supplier No. 3627104
The BeadBug 6 homogenizer uses a three-dimensional shaking motion. This motion results in high velocity impacts between the sample and the beads. This method is efficient for releasing cellular contents, such as nucleic acids and proteins. The digital display is simple to understand; program options include set speed, time, number of cycles, and pause between cycles. The last programmed parameters are stored in memory so you can easily do repeat runs.