Learn More
Roche Diagnostics BenchMark Special Stains Wash II, 1 kit
Supplier:  Roche Diagnostics 08309817001
Encompass
BenchMark Special Stains Wash II is used on the VENTANA BenchMark Special Stains automated slide stainer. When diluted according to instructions, BenchMark Special Stains Wash II produces an optimized solution for use with the VENTANA special stains reagent kits. This solution provides a stable aqueous environment for special histological staining reactions.
Catalog No. NC1859269