Learn More
Alkali Scientific BenchMasher 400 Laboratory Pad
SDP
Catalog No. 503029569
Encompass_Preferred
The BenchMasher400 Laboratory Paddle Blender IPD4400 provides fast and effective mixing ensuring consistent homogenization It is ideal for preparation of samples were eliminating contamination is critical such as pathogen and microbial detection in food and water filters The BenchMasher also has applications anywhere larger solid samples need broken down or products such as powders and polymers need mixed Samples up to 400ml are conveniently loaded in the sterile blender bags which are sealed upon closing the door Mixing paddles rapidly strike the bags producing thorough mixing and homogenization without ever directly contacting the sample Programming is user friendly mixing speed and cycle time are easily adjusted on the large full color touch screen Paddle position can be adjusted to ensure efficient processing of samples For applications requiring thermal regulation (such as Trichinella testing in meat) the chamber can be heated and maintained at temperatures up to 65C
Catalog No. 50-302-9569 Supplier Alkali Scientific Supplier No. RS2052
The BenchMasher400 Laboratory Paddle Blender IPD4400 provides fast and effective mixing ensuring consistent homogenization It is ideal for preparation of samples were eliminating contamination is critical such as pathogen and microbial detection in food and water filters The BenchMasher also has applications anywhere larger solid samples need broken down or products such as powders and polymers need mixed Samples up to 400ml are conveniently loaded in the sterile blender bags which are sealed upon closing the door Mixing paddles rapidly strike the bags producing thorough mixing and homogenization without ever directly contacting the sample Programming is user friendly mixing speed and cycle time are easily adjusted on the large full color touch screen Paddle position can be adjusted to ensure efficient processing of samples For applications requiring thermal regulation (such as Trichinella testing in meat) the chamber can be heated and maintained at temperatures up to 65C