Learn More
JDM LAB SOLUTIONS LLC BENCHMATE C12V HIGH SPEED MICR
Catalog No. NC2127780
Encompass
All centrifugation protocols can be performed up to the maximum speed of 15,000 rpm (15,595 x g) with this high speed microcentrifuge.
Catalog No. NC2127780 Supplier JDM LAB SOLUTIONS LLC Supplier No. C12V
All centrifugation protocols can be performed up to the maximum speed of 15,000 rpm (15,595 x g) with this high speed microcentrifuge.