Learn More
JDM LAB SOLUTIONS LLC Benchmate TR6 Digital Tube Roller
Supplier:  JDM LAB SOLUTIONS LLC TR6
Encompass
Speed setting from 10 to 80 rpm, and timer setting from 1 to 99 mins and infinite mode.Adjustable roller spacing to hold different diameter tubes.Programming feature – User can save and run up to 9 user-defined programs (protocols).Microprocessor controlled for accurate speed under variable loads.Pulse mode for more vigorous mixing (alternating rotating direction). The user can set the pulse time from 30 to 99 seconds.Stackable design with robust construction.
Catalog No. NC2385837