Learn More
Alkali Scientific Alkali Scientific™ BenchMixer™ MP Vortex Mixer (BV1007) for Tubes and Plates
SDP
Catalog No. 503029652
Encompass_Preferred
The BenchMixer MP (BV1007) is optimally designed to provide thorough mixing in a variety of commonly used sample vessels. With included holders for tubes and plates, as well as a cup head for pressurized/touch vortexing, the BenchMixer MP is a complete package designed to meet the mixing needs of any laboratory. Mixing speed is adjustable from 200 to 3,000 rpm and time can be set up to 999 minutes or continuous. A unique, automatic recognition feature detects the installed sample holder and adjust the maximum speed setting accordingly. With a compact footprint (only 5 inches wide) and powerful suction cup feet, the mixer conserves bench space and ensures stability during mixing. Accepts a wide range of microplates, including deep well, PCR, Elisa plates, etc. Includes tube accessories for vortexing of 1.5/2.0 ml,15 ml, or 50 ml tubes Converts to a traditional “touch” mode vortexer with included cup head attachment Powerful vortexing at speeds up to 3,000 rpm
Catalog No. 50-302-9652 Supplier Alkali Scientific Supplier No. RS1007
The BenchMixer MP (BV1007) is optimally designed to provide thorough mixing in a variety of commonly used sample vessels. With included holders for tubes and plates, as well as a cup head for pressurized/touch vortexing, the BenchMixer MP is a complete package designed to meet the mixing needs of any laboratory. Mixing speed is adjustable from 200 to 3,000 rpm and time can be set up to 999 minutes or continuous. A unique, automatic recognition feature detects the installed sample holder and adjust the maximum speed setting accordingly. With a compact footprint (only 5 inches wide) and powerful suction cup feet, the mixer conserves bench space and ensures stability during mixing. Accepts a wide range of microplates, including deep well, PCR, Elisa plates, etc. Includes tube accessories for vortexing of 1.5/2.0 ml,15 ml, or 50 ml tubes Converts to a traditional “touch” mode vortexer with included cup head attachment Powerful vortexing at speeds up to 3,000 rpm