Learn More
Research Products International Corp BenchMixer Vortex Mixer with Q-Drive Dynamic Balance System
SDP
Catalog No. 501367756
Encompass
BenchMixer Vortex Mixer features a Q-Drive dynamic balance system that provides extremely low vibration, quiet operation and long motor life. Vortex Mixer provides smooth instant vortexing of even the largest sample sizes, including 50 ml tubes. Additional features include Touch or Continuous Operation, no-walk suction feet, cold room/incubator compatibility and spill proof electronic controls.
Catalog No. 50-136-7756 Supplier Research Products International Corp Supplier No. 400810
BenchMixer Vortex Mixer features a Q-Drive dynamic balance system that provides extremely low vibration, quiet operation and long motor life. Vortex Mixer provides smooth instant vortexing of even the largest sample sizes, including 50 ml tubes. Additional features include Touch or Continuous Operation, no-walk suction feet, cold room/incubator compatibility and spill proof electronic controls.