Learn More
Braintree Scientific Benchmixer Vortex Mixer combo head for tubes & microplates
SDP
Catalog No. 501954458
Encompass
Benchmixer sets the standard in vortex mixing. With advanced Q-Drive™technology, BenchMixer™ provides smooth instant vortexing of even the largest sample sizes, including full 50ml tubes. Unique counter balance system creates maximum vortexing action, while minimizing noise and excessive vibration resulting in quieter, more efficient and longer lasting operation. Other features include: "no-walk" suction feet, cold room/incubator compatibility and spill-proof electronic controls. Powerful Motor for instant vortexing, Q-Drive™ Dynamic Balance System, Touch or Continuous Operation, Variable Speed Control from 200 to 3200 rpm. *Note: Benchmixer is not included (see part# 501954457 for mixer) **Sold for research purposes only and not for human use**
Catalog No. 50-195-4458 Supplier Braintree Scientific Supplier No. BV1000COM
Benchmixer sets the standard in vortex mixing. With advanced Q-Drive™technology, BenchMixer™ provides smooth instant vortexing of even the largest sample sizes, including full 50ml tubes. Unique counter balance system creates maximum vortexing action, while minimizing noise and excessive vibration resulting in quieter, more efficient and longer lasting operation. Other features include: "no-walk" suction feet, cold room/incubator compatibility and spill-proof electronic controls. Powerful Motor for instant vortexing, Q-Drive™ Dynamic Balance System, Touch or Continuous Operation, Variable Speed Control from 200 to 3200 rpm. *Note: Benchmixer is not included (see part# 501954457 for mixer) **Sold for research purposes only and not for human use**