Learn More
Chemglass Life Sciences BenchMixer Vortexer 115V
SDP
Catalog No. NC1378858
Encompass
BenchMixer Vortexer 115V. The BenchMixer™ sets the new standard in vortex mixing. With advanced Q-Drive™ technology, the Benchmark BenchMixer provides smooth instant vortexing of even the largest sample sizes, including full 50ml tubes. The unique counter-balance system creates maximum vortexing action, while minimizing noise and excessive vibration. This results in quieter, more efficient and longer lasting operation.
Catalog No. NC1378858 Supplier Chemglass Life Sciences Supplier No. CLS4890A01
BenchMixer Vortexer 115V. The BenchMixer™ sets the new standard in vortex mixing. With advanced Q-Drive™ technology, the Benchmark BenchMixer provides smooth instant vortexing of even the largest sample sizes, including full 50ml tubes. The unique counter-balance system creates maximum vortexing action, while minimizing noise and excessive vibration. This results in quieter, more efficient and longer lasting operation.