Learn More
Sigma Aldrich Fine Chemicals Biosciences BenchMixerTM Vortexer AC/D1EA
Catalog No. 502905286
Encompass
The Mortexer is MORE than your standard vortex mixer. The unique design of the Multi-Head (included) features a traditional cup head for general purpose mixing plus holds up to eight microcentrifuge tubes.The drive system is balanced with Q-Drive technology providing a smooth vortexing action while minimizing noise and excessive vibration. This results in quieter more efficient and longer lasting operation. Increased speed up to 3400 rpm produces an instant vortex of even the largest sample sizes including full 50ml tubes.Other features include no-walk suction feet cold room/incubator compatibility and spill-proof electronic controls.Dimensions (WxDxH) 5 x 6.3 x 6.7 in. or 13 x 16 x 17 cmWeight 8.4 lbs (3.8 kg)Operating modes Touch or continuuous
Catalog No. 50-290-5286 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. BMSBV10001EA
The Mortexer is MORE than your standard vortex mixer. The unique design of the Multi-Head (included) features a traditional cup head for general purpose mixing plus holds up to eight microcentrifuge tubes.The drive system is balanced with Q-Drive technology providing a smooth vortexing action while minimizing noise and excessive vibration. This results in quieter more efficient and longer lasting operation. Increased speed up to 3400 rpm produces an instant vortex of even the largest sample sizes including full 50ml tubes.Other features include no-walk suction feet cold room/incubator compatibility and spill-proof electronic controls.Dimensions (WxDxH) 5 x 6.3 x 6.7 in. or 13 x 16 x 17 cmWeight 8.4 lbs (3.8 kg)Operating modes Touch or continuuous