Learn More
Braintree Scientific BenchRocker 2D variable speed rocker platform elastic tie
SDP
Catalog No. 501954354
Encompass
Benchrocker With adjustable speeds and tilt angles, user can set both parameters to match vessel size and volume of liquid being mixed. User can adjust both parameters to match vessel size and volume of liquid being mixed, yielding optimum results. Rocking speed and tilt angle are both easily adjustable in seconds. At low speeds and minimal tilt angles, both models (2D for "see-saw" motion or 3D for "gyratory" motion) provide a gentle wave effect for non-foaming mixing, as is required for most blot washing and gel work. When set to higher speeds and more extreme tilt angles, they produce aggressive agitation for vigorous sample mixing. Dual platform option expands workspace. BenchRocker 2D and 3D can accept stackable platforms (2.75" clearance). All platform mats are autoclavable. Both models have maintenance free brushless motors and are safe for use in cold rooms and incubators. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4354 Supplier Braintree Scientific Supplier No. BR2000B0718
Benchrocker With adjustable speeds and tilt angles, user can set both parameters to match vessel size and volume of liquid being mixed. User can adjust both parameters to match vessel size and volume of liquid being mixed, yielding optimum results. Rocking speed and tilt angle are both easily adjustable in seconds. At low speeds and minimal tilt angles, both models (2D for "see-saw" motion or 3D for "gyratory" motion) provide a gentle wave effect for non-foaming mixing, as is required for most blot washing and gel work. When set to higher speeds and more extreme tilt angles, they produce aggressive agitation for vigorous sample mixing. Dual platform option expands workspace. BenchRocker 2D and 3D can accept stackable platforms (2.75" clearance). All platform mats are autoclavable. Both models have maintenance free brushless motors and are safe for use in cold rooms and incubators. **Sold for research purposes only and not for human use**