Learn More
Sigma Aldrich Fine Chemicals Biosciences BenchRockerTM variable 2D1EA
Catalog No. 502908895
Encompass
The BenchRocker 2D is designed to provide the precise speeds and tilt angles required for a broad range of molecular and biological mixing applications. The user can adjust both parameters to match the vessel size and the volume of liquid being mixed yielding optimum results.
Catalog No. 50-290-8895 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7554781EA
The BenchRocker 2D is designed to provide the precise speeds and tilt angles required for a broad range of molecular and biological mixing applications. The user can adjust both parameters to match the vessel size and the volume of liquid being mixed yielding optimum results.