Learn More
Thomas Scientific Absorbent Liner, 20" x 300', 1 Roll
SDP
Catalog No. 501458307
Encompass
Protects surfaces and absorbs liquids Protect benchtops, floors and carts with this highly absorbent cover. Keeps spills from spreading. Cushions the surface while absorbing liquids. Backing is waterproof and resistant to most chemicals. Made of highly absorbent three-ply tissue with a polypropylene backing. Unique Features Economical Absorbs spills Retains liquids Soft and durable Can be cut to size EZROLL Absorbent Liner Dispenser Rack Fits any roll up to 26" width and 20" diameter. Mount on any secure surface. Heavy duty steel cnstruction. Fits perfectly with Jaece Absorbent Liner Rolls.
Catalog No. 50-145-8307 Supplier Thomas Scientific Supplier No. 1201T88
Protects surfaces and absorbs liquids Protect benchtops, floors and carts with this highly absorbent cover. Keeps spills from spreading. Cushions the surface while absorbing liquids. Backing is waterproof and resistant to most chemicals. Made of highly absorbent three-ply tissue with a polypropylene backing. Unique Features Economical Absorbs spills Retains liquids Soft and durable Can be cut to size EZROLL Absorbent Liner Dispenser Rack Fits any roll up to 26" width and 20" diameter. Mount on any secure surface. Heavy duty steel cnstruction. Fits perfectly with Jaece Absorbent Liner Rolls.