Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kinematica Inc Benchtop Autoclave Sterilizer

Catalog No. 12983545 Shop All Kinematica Products
Change view
Click to view available options
Capacity (Metric):
3 L
30 L
56 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
12-983-545 56 L
12-983-543 30 L
12-983-544 3 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12-983-545 Supplier Kinematica Inc Supplier No. PRIME56L
Add to Cart
Add to Cart
Request a Quote

Diversified Applications : Agar Preparation, Liquid Sterilization, Empty Glassware Sterilization, Instrument and Solid Sterilization, Waste Liquid / Solid / Mixed Sterilization, Respirator Mask Sterilization,Other Heat Resistant Materials.

  • Features:
  • Advanced Steri-Touch Panel HMI
  • High-Accuracy Temperature Control
  • Integrated Steam generator
  • Flexible Load and Personnel Management
  • Powerful Vacuum System
  • Comprehensive Filtration System
  • Fully Customizable Programs
  • F0 Value Calculation
  • Includes Helix
  • Bowie-Dick
  • Ample 2G Data Storage
  • Automatic Water Management
  • Integrated Digital Recorder
  • Thermal Printer

Specifications

Autoclavable Autoclavable
Capacity (English) 14.7 gal.
Capacity (Metric) 56 L
Depth (Metric) Chamber 110 mm
Diameter (English) Chamber 16 in.
Depth (English) Chamber 4.3 in.
Diameter (Metric) Chamber 380 mm
Length (English) 31.5 in.
Length (Metric) 800 mm
Product Type Benchtop Autoclave
Temperature (English) 249°F to 275°F
Temperature (Metric) 121°C to 134°C
Latex Free Yes
Sterility Sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.