Learn More
Research Products International Corp Benchtop BioHazard Bag Holder, Small
SDP
Catalog No. 50998025
Encompass
Ideal system for keeping disposal bags open and ready to use. Eliminates having to remove lids from containers to dispose contaminated materials. Stainless steel wire frame and sturdy polypropylene base hold bag securely in place. Entire unit can be autoclaved. Assembles in minutes and is permanent.
Catalog No. 50-998-025 Supplier Research Products International Corp Supplier No. 246100
Ideal system for keeping disposal bags open and ready to use. Eliminates having to remove lids from containers to dispose contaminated materials. Stainless steel wire frame and sturdy polypropylene base hold bag securely in place. Entire unit can be autoclaved. Assembles in minutes and is permanent.