Learn More
Research Products International Corp Benchtop BioHazard Bag Holder, Large
SDP
Catalog No. 501977963
Encompass
Ideal system for keeping disposal bags open and ready to use. Eliminates having to remove lids from containers to dispose contaminated materials. Stainless steel wire frame and sturdy polypropylene base hold bag securely in place. Entire unit can be autoclaved. Assembles in minutes and is permanent.
Catalog No. 50-197-7963 Supplier Research Products International Corp Supplier No. 246102
Ideal system for keeping disposal bags open and ready to use. Eliminates having to remove lids from containers to dispose contaminated materials. Stainless steel wire frame and sturdy polypropylene base hold bag securely in place. Entire unit can be autoclaved. Assembles in minutes and is permanent.