Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Benchtop Biohazard Disposal Can with Motion Sensor Lid
SCI_ED

Catalog No. S72309 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Capacity (Metric):
1.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S72309 1.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S72309 Supplier Bel-Art™ Supplier No. H131941011/EMD
  • $1,141.00
    $839.65 / Case of 12

    Save $301.35

    26% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Compact benchtop biohazard disposal can contains odors and is perfect for small items such as pipette tips and microcentrifuge tubes.

  • Automatic opening lid provides hands free use; closes automatically and securely for a more sanitary environment
  • Holds a 25 x 25 cm (10 x 10 in.) autoclavable biohazard disposal bag (S43506)
  • Includes a polypropylene liner with handle for easy emptying of contents
  • Aesthetic and durable; stainless steel body and chrome lid are resistant to bleach and easy to clean
  • Requires 3 AAA batteries (not included)
  • Compliance: CE & RoHS

Specifications

Capacity (Metric) 1.5 L
Certifications/Compliance CE and RoHS
Closure Type Chrome Lid
Diameter (Metric) Outer 12.4 cm
Diameter (English) Outer 4.8 in.
Capacity (English) 0.4 gal.
Color Silver
Height (Metric) 17 cm
Autoclavable Not Autoclavable
Includes Polypropylene liner with handle
Liner Material Polypropylene
For Use With (Application) Containing Odors; Sanitary Waste Disposal
For Use With (Equipment) 10 x 10 in. autoclavable biohazard disposal bag (H13182-0000)
Height (English) 6.7 in.
Material Stainless Steel
Product Type Disposal Can
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.