Learn More
Azzota Corp Benchtop Conductivity Meter With Temp Conductivity Range: 0.05 - 2X105 Us/Cm Temp Range: 0-100Deg.C Temp Compensation: 0-40Deg.C Temp: Coefficient: 0-5% Per Deg.C Dimension: 210mm X 205mm X 65mm
Catalog No. 507532965
Encompass_Preferred
Benchtop Conductivity Meter with Temperature Conductivity Range: 0.05 - 2x105 uS/cm Temperature Range: 0-100deg.C Temperature Compensation: 0-40deg.C Temperature: Coefficient: 0-5% per deg.C Dimension: 210mm x 205mm x 65mm
Catalog No. 50-753-2965 Supplier Azzota Corp Supplier No. CM10T
Benchtop Conductivity Meter with Temperature Conductivity Range: 0.05 - 2x105 uS/cm Temperature Range: 0-100deg.C Temperature Compensation: 0-40deg.C Temperature: Coefficient: 0-5% per deg.C Dimension: 210mm x 205mm x 65mm