Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Benchtop Cooler

Catalog No. 14127636 Shop All Fisherbrand Products
Change view
Click to view available options
Array:
3 x 4
4 x 8
Color:
Blue
Green
Includes:
Non-Insulated Lid
PCM Insulated Lid
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Array Color Includes
14-127-636 3 x 4 Green Non-Insulated Lid
14-127-647 3 x 4 Blue Non-Insulated Lid
14-127-656 4 x 8 Green Non-Insulated Lid
14-127-646 4 x 8 Blue Non-Insulated Lid
14-127-558 4 x 8 Green PCM Insulated Lid
14-127-634 4 x 8 Blue PCM Insulated Lid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 14-127-636
Add to Cart
Add to Cart

Compact cooling designed for accuracy, used on the lab bench to keep samples and reagents cold

  • The Mini Cooler protects temperature-sensitive samples at the bench or in the freezer with the Mini Cooler.
  • They are ideal for keeping PCR reagents, enzymes, and other biochemical reagents cool while working at the bench.

Specifications

Height (Metric) 8.4 cm
Length (Metric) 14.2 cm
Width (Metric) 10.2 cm
Dimensions (L x W x H) 5.6 x 4 x 3.3 in.
Includes Non-Insulated Lid
Array 3 x 4
Material Polycarbonate
Type Mini Cooler
Color Green
Capacity (Metric) 2 mL
Holds 12 Tubes (0.5 to 2 mL)
Autoclavable Not Autoclavable
Temperature Control Passive
Temperature Range -30°C to +40°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.