Learn More
Fisherbrand™ Benchtop Extraction Kit
Catalog No. 15050086 Shop All Fisherbrand Products
Change view
Click to view available options
Product Type:
Carbon Extraction Kit
Carbon Filter
Filter Connector
Standard Extraction Kit
Standard Filter
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
15-050-086 Standard Extraction Kit
15-050-087 Carbon Extraction Kit
15-050-088 Standard Filter
15-050-089 Carbon Filter
15-050-090 Filter Connector
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 15-050-086
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
Benchtop Extraction Kits used for the capture and containment of particulate generated on the bench space.
  • The Benchtop extraction kits include arms with hood or nozzle, fan with speed control and standard filter (particle, HEPA or gas), hose, connection details, and table bracket.
  • The speed of the fan is variably controlled by a speed controller.
  • Complete with hoses and extraction hoods.
  • The hood is made of clear plastic, which increases the visibility.
Specifications
Product Type Standard Extraction Kit
Color White
Material Aluminum