Learn More
Yamato Scientific America, Inc. Yamato 37L Incubator with window, General Purpose, Benchtop, 220V
Catalog No. 501898081 Shop All Yamato Scientific America Inc. Products
Encompass_Preferred
Benchtop, economical general purpose 37L natural convection incubator with observation window. Control panel located at a higher position for easy access. Easy to use digital setting display and timer. Air jacket technology ensures even and efficient heat distribution throughout the chamber. Standard equipped with various functions such as self-diagnostic, calibration off-set, overheat prevention and key lock. Operating temperature range: +5~ 80°C
Catalog No. 50-189-8081 Supplier Yamato Scientific America, Inc. Supplier No. IC113CW
Benchtop, economical general purpose 37L natural convection incubator with observation window. Control panel located at a higher position for easy access. Easy to use digital setting display and timer. Air jacket technology ensures even and efficient heat distribution throughout the chamber. Standard equipped with various functions such as self-diagnostic, calibration off-set, overheat prevention and key lock. Operating temperature range: +5~ 80°C