Learn More
Description
- The Liquid Handling Station Flow (LHS Flow) are compact benchtop automated liquid handling systems designed to simplify repetitive pipetting workflows while improving consistency and efficiency.
- Intuitive software enables users to create and modify methods quickly, without requiring programming experience, making automation accessible for a wide range of laboratory applications.
- Key Features and Benefits:Automates routine pipetting tasks, freeing laboratory personnel for Higher-value work User-friendly software allows method creation in minutes with no programming experience required compact benchtop footprint fits easily into Space-constrained laboratories quiet operation with adjustable settings to accommodate various workflows.
- Cost-effective automation solution for Small- to Medium-volume sample processing reduces manual pipetting errors and improves reproducibility across experiments flexible worktable configuration supports a variety of labware formats and applications responsive technical support, installation assistance, and training available.
- Performance Specifications:Single-channel volume ranges: 150 L, 10200 L, 401000 L8-channel volume ranges: 150 L, 20300 L, 401000 L working positions: Seven configurable work positions (P2P8) plus P1 dedicated for waste disposal Applications:PCR and qPCR setup ELISA preparation enzyme assays serial dilutions cherry picking plate replication aliquoting general sample preparation.
- Versatile design:Seven configurable working positions support microplates (24-, 96-, and 384-well), tubes, vials, reservoirs, and other laboratory vessels supports diverse assay and sample preparation workflows easily adapts to changing laboratory requirements compact Footprint:Approximate dimensions with door closed (Width x Depth x Height): LHS: 24 x 20 x 21 in.; LHS Flow: 24 x 21 x 27 in. Approximate dimensions with door open (Width x Depth x Height): LHS: 24 x 20 x 28 in.; LHS Flow: 24 x 21 x 28 in.
- One of the smallest footprints in its class fits conveniently on laboratory benches and under overhead cabinets available configuration for applications requiring a controlled environment, the LHS Flow model incorporates a built-in horizontal laminar flow enclosure to help maintain contamination-free liquid handling conditions.
- The system is designed to meet ISO 14644-1 Class 5 and GMP Annex 1 Class A requirements.Designed for reliability, flexibility, and ease of use, the LHS and LHS Flow systems help laboratories improve productivity, enhance reproducibility, and streamline everyday liquid handling workflows.
- Automate repetitive pipetting tasks with intuitive software, flexible configurations, and a compact benchtop footprint.
Specifications
Specifications
| Product Type | Automated Pipetting System |
| Channels | 1, 8 |
| Material | Stainless Steel |
| Volume (Metric) | 152.93 L |
| For Use With (Application) | Automated Liquid Handling and Pipetting |
| For Use With (Equipment) | LHS and LHS Flow Systems |
| Includes | Motor Control Unit, Operating Software, Operating Manual, Power Cable, USB Cable, Documentation, and On-site Training |
| Interface | 1 x USB |
| Sterility | Non-sterile |
| Dimensions (L x W x H) | 48.5 x 59.5 x 53 cm (19.09 x 23.43 x 20.87 in.) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.