Learn More
Electron Microscopy Sciences Bench Top Mat 50 FT
SDP
Catalog No. 5024667
Encompass_Preferred
A flexible polyethylene film backing which has a highly absorbent upper surface which traps water, dyes, resins, solvents, and radioactive solutions. The sheets can be rolled up and discarded after use. Ideal for daily use in EM labs. Roll 20" wide and 50 ft. long in a dispenser carton
Catalog No. 50-246-67 Supplier Electron Microscopy Sciences Supplier No. 64445
A flexible polyethylene film backing which has a highly absorbent upper surface which traps water, dyes, resins, solvents, and radioactive solutions. The sheets can be rolled up and discarded after use. Ideal for daily use in EM labs. Roll 20" wide and 50 ft. long in a dispenser carton